Nitric acid 65%, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Nitric acid solution
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:87
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Molecular Weight:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8 (5.1) II
Merck Index:13,06610
:M-Clarity™ quality level = MQ100
:GHS03
:GHS06
:GHS05
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-32 °C
- Boiling Point:121 °C (1013 hPa)
- Vapor Pressure:9.4 hPa (20 °C)
- Density:1.39 g/cm3 (20 °C)
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00441.0250_TDS_EN MERC1.00441.1000_TDS_EN ProductPage_100441 ProductPage_100441
Product specification
Merck Pharma Segment Brochure Merck Ultrapur and Suprapur® High Purity Acids and Bases
Product Literature
msds_100441
SDS
Frequently Bought Together