Order Entry
Denmark
ContactUsLinkComponent
Heptane (mixture of isomers), TECHNICAL
Heptane (mixture of isomers), TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Heptane (mixture of isomers), TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    min. 98.5%
  • Refractive index (20°C):
    1.39 - 1.405
  • Evaporation residue:
    Max. 20 ppm
  • Boiling point:
    90 - 100 °C

Specification Test Results

Frequently Bought Together