Order Entry
Denmark
ContactUsLinkComponent
 
(-)-Quinine ≥99.0% for racemat separation, for synthesis, Sigma-Aldrich®
  8.02304.0025
 :  Merck
 :  
(-)-Quinine ≥99.0% for racemat separation, for synthesis, Sigma-Aldrich®
  8.02304.0025
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

 

 

  • Assay (HClO₄):
    ≥ 99,0 %(m)
  • Melting range (lower value):
    ≥ 174 °C
  • Melting range (upper value):
    ≤ 178 °C
  • Spec. rotation [α²0/D (c=1 in H₂SO₄ 0.1 mol/l):
    -285,0 - -275,0 °
  • Identity (IR):
    conforms

 

 

 : (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
 :
 :324.42 g/mol
 :205-003-2
 :GHS08
 :Ambient
 :130-95-0
 :C₂₀H₂₄N₂O₂
 :MFCD00198096
 :176
 :12,08245
 :M-Clarity™ quality level = MQ200