Order Entry
Denmark
ContactUsLinkComponent
Vortex mixers, Vortex-Genie®
undefined
Vortex mixers, Vortex-Genie®
  • Catalog No:
  • 444-9173
  • 444-9174
  • 444-5900
  • Model:
  • Vortex-Genie® 1
  • Vortex-Genie® 2T
  • Vortex-Genie® 2
  • Shaking motion:
  • Orbital
  • Orbital
  • Orbital
  • Orbit:
  • 4 mm
  • 4 mm
  • 4 mm
  • Speed range:
  • 2700 min⁻¹ fixed
  • 600 - 2700 min⁻¹
  • 600 - 2700 min⁻¹
  • Max. capacity:
  • 500 ml*
  • 500 ml*
  • 500 ml*
  • W×D×H:
  • 122×165×165 mm
  • 122×165×165 mm
  • 122×165×165 mm
  • Weight:
  • 4 kg
  • 4 kg
  • 4 kg
Frequently Bought Together