Order Entry
Denmark
ContactUsLinkComponent
System for testing jet fuel, HY-LiTE® Jet A1 Fuel Test
System for testing jet fuel, HY-LiTE® Jet A1 Fuel Test
Catalog # 1.30196.0021
Supplier:  Merck
undefined
System for testing jet fuel, HY-LiTE® Jet A1 Fuel Test
Catalog # 1.30196.0021
Supplier:  Merck
Specifications
  • Description:
    HY-LiTE® Jet A1 fuel test kit
  • Environmentally Preferable:
  • Pk:
    20 Tests
Specifications
About this item
For aviation fuel used in civilian air transport. Aviation fuel contains small quantities of water, so it is vulnerable to microbiological attack. Microbiological contamination in fuel can lead to operational problems, e.g. clogged filters or corroded tanks. This test is an alternative for anyone who monitors or deals with aviation fuel. Previously, test results were only available after three days. With the HY-LiTE® test, results are returned in just 10 minutes. This test is recommended in the IATA (International Air Transport Association) Directive.
  • A simple, on-site test, documentation in the device
  • Same method for samples with or without water
  • Shows biological activity directly in the sample, regardless of bacteria or other bio films
  • Quantitative results and simple interpretation
  • Flexible test volume (1 L is recommended)
Delivery: Kit includes 20 pens plus 20 long and 20 short pipettes.