Ceran® hotplates, LHG
Supplier: GESTIGKEIT HARRY
Certificates
About this item
Glass ceramic heating plate, which is extremely resistant to breakage and changes of temperature, will not distort, is permeable to ultra-violet light and also highly acid-resistant.
- Uniform heating across entire surface
- Insulated stainless steel housing
- Temperature control by an NiCr-Ni sensor (regulation from 50 to 500 °C)
Four feet, individually adjustable, ensure wobble-free, horizontal stability.
Caution: Note: models 33 A/SR and 44 A/SR requires a 3-phase (400 V) supply.
Frequently Bought Together