Zero air generators
Designed as a compact, hazard free, lower cost alternative to the user of expensive high pressure gas cylinders in the laboratory. Replacing cylinders with a gas generator brings advantages: increased safety, greater convenience, reduced cost and higher purity gas for more accurate analysis.
- Produces zero air from compressed air
- Ensures a flat and consistent base line
- Short payback period
- Compact and reliable
Purity: <0,05 ppm THC.
Inlet pressure 2,8 to 8,6 bar.
Inlet/outlet port size: ¼" NPT female.