Strips of 8 PCR tubes with attached flat individual caps
Supplier: Brand
Certificates
About this item
PP. Eight connected 0,2 or 0,15 ml PCR tubes with attached, flat individual caps.
- Cap offers protection against contamination
- Highly transparent cap makes them ideal for real-time PCR (qPCR)
- Available in standard or low profile versions
- Free from DNase, RNase and DNA
Colour: Clear