Stoppers, rubber, conical, with holes
Conical natural grey rubber stoppers with one hole.
- Resistant to common laboratory chemicals
- Highly elastic, for a perfect seal and exceptional bonding
- With pre-drilled holes
- Withstands temperatures from −25 to +70 °C (+120 °C in humid atmosphere)
- Hardness (shore durometer A): 40
Withstands temperatures from -25 to +70 °C (+120 °C in moist heat)
Hardness (Shore durometer A): 40
DIN 12871
: DIN 12871
Frequently Bought Together