Order Entry
Denmark
ContactUsLinkComponent
Spatula set
  231-2176
 :  RSG
undefined
Spatula set
  231-2176
 :  RSG
  • Description:
    Set of 6 spatulas and 1 pair of tweezers
  • Environmentally Preferable:
  • Pk:
    1 SET
Stainless steel 18/10. Useful combination of double spatulas, spoon spatulas and forceps. Universal usage in laboratories and industry.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Comes in practical imitation leather case
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as tweezers (straight, blunt, 145 mm long).
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as forceps (straight, blunt, 145 mm long).