Order Entry
Denmark
ContactUsLinkComponent
Tongue depressor
Tongue depressor
Catalog # HECH44365020
undefined
Tongue depressor
Catalog # HECH44365020

Specifications

  • Width:
    18 mm
  • Description:
    Tongue depressor, non sterile, in small box
  • Length:
    150 mm
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    100

Specifications

About this item

Wood.

  • Smooth, round edges
  • Clean finish, splinter-free
  • Odour- and taste-free
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904