Pipette controller, PIPETBOY acu 2
: Integra
Pipette controller for glass and plastic pipettes from 0,1 to 100 ml.
- Lightweight (195 g), enhanced ergonomics for stress and fatigue-free pipetting
- Pipetting speed is conveniently pre-set via thumb wheel; speed is then easily regulated with the tips of the fingers
- Autoclavable nose piece and easy to replace hydrophobic filter for reliable protection
- LED provides clear information about the battery charging status
- Colour coding for easy identification of the right controller
- Maximum pipetting speed: 13,5 ml/s (with 50 ml pipette)
The second generation of PIPETBOY acu is equipped with the latest technology providing increased pipetting speed and significant instrument weight reduction. These new features, combined with unchanged main benefits, provide an improved ergonomic pipetting experience.
:
Supplied with a rechargeable Li-ion battery, wall holder, mains adapter and 0,45 µm sterile filter. Additional filters (0,2 and 0,45 µm) can be ordered separately.
Frequently Bought Together