Order Entry
Denmark
ContactUsLinkComponent
Glove box, controlled atmosphere, Protector®
Glove box, controlled atmosphere, Protector®
Supplier:  LABCONCO
Certificates
undefined
Glove box, controlled atmosphere, Protector®
Supplier:  LABCONCO

Specifications

  • Catalog No:
  • LBCP5080035
  • LBCP5080031
  • LBCP5060035
  • Model:
  • Protector®
  • Protector®
  • Protector®
  • Observation window (W×H):
  • 914,5×701 mm
  • 914,5×701 mm
  • 914,5×701 mm
  • Glove port - int.Ø:
  • 203 mm
  • 203 mm
  • 203 mm
  • W×D×H ext.:
  • 1623×874×1161 mm
  • 1623×874×1161 mm
  • 1623×874×1161 mm
  • W×D×H int.:
  • 889×873×1160 mm
  • 889×873×1160 mm
  • 889×873×1160 mm
  • Weight:
  • 227 kg
  • 227 kg
  • 227 kg

Specifications

Related Information

Frequently Bought Together