About this item
The Visualizer Western Blot Detection kit is designed for the sensitive detection of proteins in chemiluminescent Western blots.
Storage: Stable for 18 months at 4 °C
- Application: For use on immunoblots
The reagents in this kit have been matched to optimise the range and sensitivity of detection using various antibodies in Western blot assays.
Components: Goat anti-mouse IgG, goat anti-rabbit IgG, HRP conjugate Visualizer detection reagent A Visualizer detection reagent B.
Visualizer EC western blot detection kits are designed with a high glow reaction chemiluminescent substrate with outstanding sensitivity, long-lasting signal and low cost. Visualizer EC offers the highest signal-to-noise ratios of any existing luminol peroxidase formulations, resulting in high sensitivity and low backgrounds.
Specifications
- Environmentally Preferable: