Order Entry
Denmark
ContactUsLinkComponent
Methyl-10-undecenoate 97%
Methyl-10-undecenoate 97%
New Product
Certificates
Methyl-10-undecenoate 97%
New Product

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    clear colorless to yellow liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96 %
  • Refractive index:
    1.4380 to 1.4400 (20°C, 589 nm)

Specification Test Results

Frequently Bought Together