X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) enzyme substrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
CAS Number:7240-90-6
:
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666
Molecular Weight:408.63 g/mol
Melting Pt:237…239 °C
Storage Temperature:Freezer
EINECS:230-640-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together