Order Entry
Denmark
ContactUsLinkComponent
 
Anti-SOX11 Mouse Monoclonal Antibody [clone: SOX11/7236]
Catalog # ANTIA277801-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-SOX11 Mouse Monoclonal Antibody [clone: SOX11/7236]
Catalog # ANTIA277801-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MRD27
  • Antigen symbol:
    SOX11
  • Clonality:
    Monoclonal
  • Clone:
    SOX11/7236
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P35716
  • Antigen synonyms:
    SRY (sex determining region Y) box 11|SOX11|SRY-box 11|SRY related HMG box gene 11|Transcription factor SOX-11|SOX11_HUMAN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    47 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 50-200, of human SOX11 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [SOX11/7236] antibody to SOX11 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: SOX11
Clonality: Monoclonal
Clone: SOX11/7236
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human