Order Entry
Denmark
ContactUsLinkComponent
 
Anti-Galectin 3 Mouse Recombinant Antibody [clone: rLGALS3/7286]
  ANTIA314989-100
New Product
Anti-Galectin 3 Mouse Recombinant Antibody [clone: rLGALS3/7286]
  ANTIA314989-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    35 kDa lectin
  • Antigen symbol:
    Galectin 3
  • Clonality:
    Recombinant
  • Clone:
    rLGALS3/7286
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17931
  • Antigen synonyms:
    Carbohydrate-binding protein 35|Galactose-specific lectin 3|GAL3|CBP35|GALBP|CBP 35|Carbohydrate binding protein 35|Gal-3|Galactoside-binding protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    30 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment around LGALS3 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rLGALS3/7286] antibody to Galectin 3 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Galectin 3
Clonality: Recombinant
Clone: rLGALS3/7286
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human