Anti-HMCES Rabbit Polyclonal Antibody
Catalog # PRSI55-547
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein
- Antigen symbol:HMCES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino Acid Range = 281-310
- Form:Liquid
- Gene ID:Q96FZ2
- Antigen synonyms:HMCES
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:41 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DC12 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 281-310 amino acids from the C-terminal region of human DC12.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: HMCES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human