Anti-B4GALT3 Rabbit Polyclonal Antibody
Catalog # PRSI16-988
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Beta-1,4-galactosyltransferase 3
- Antigen symbol:B4GALT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:8703
- Antigen synonyms:epididymis secretory sperm binding protein|polypeptide 3|N-acetyllactosamine synthase|nal synthase|beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3|beta-1,4-galactosyltransferase 3|beta-1,4-GalTase 3|beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase|UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3|UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase|UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3|beta4Gal-T3|b4Gal-T3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 40-150 of human B4GALT3 (NP_003770,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: B4GALT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat