Specifications
- Antibody type:Primary
- Antigen name:Potassium voltage-gated channel subfamily V member 2
- Antigen symbol:KCNV2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino acid range = 478-507
- Form:Liquid
- Gene ID:Q8TDN2
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:62 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This KCNV2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 478-507 amino acids from the C-terminal region of human KCNV2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium voltage-gated channel subfamily V. This member is identified as a 'silent subunit', and it does not form homomultimers, but forms heteromultimers with several other subfamily members. Through obligatory heteromerization, it exerts a function-altering effect on other potassium channel subunits. This protein is strongly expressed in pancreas and has a weaker expression in several other tissues.
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: KCNV2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human