Anti-HYLS1 Rabbit Polyclonal Antibody
Catalog # PRSI23-489
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:HYLS1 centriolar and ciliogenesis associated
- Antigen symbol:HYLS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:219844
- Antigen synonyms:HLS|hydrolethalus syndrome protein 1|hydrolethalus syndrome 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 70-299 of human HYLS1 (NP_659451.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: HYLS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human