Hydrochloric acid 4 M HCl in water, Fluka™
Catalog # 84435-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (color):complying
- Appearance (form):complying
- Assay:3.95 - 4.10 mol/l
Specification Test Results
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
:
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Density:1.072 g/cm³ (20 °C)
ADR:8 II