(±)-3-Hydroxybutyric acid 95%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid
CAS Number:300-85-6
:
Formula:C₄H₈O₃
MDL Number:MFCD00004546
Boiling Pt:118…120 °C (2 mmHg)
Molecular Weight:104.11 g/mol
Flash Pt:112 °C
Merck Index:12,04861
EINECS:206-099-9
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to white to light yellow Solid or liquid
- Infrared spectrum:Conforms
- Titration with NaOH:94 to 106 %
- HPLC:≥98.5 %
- Specific optical rotation:+80° to +86° (20°C, 589 nm) (c=1, DMF)
Specification Test Results
Frequently Bought Together