Call for surface tension test
765-0136
- Description:Call for surface tension test
- Environmentally Preferable:
- Pk:1
Aluminum ring with suspension in three points. Used together with a dynamometer to determine the surface tension in liquids.
- Diameter: 65 mm