Order Entry
Denmark
ContactUsLinkComponent
Electrostatic motor
Electrostatic motor
Catalog # 765-0186
undefined
Electrostatic motor
Catalog # 765-0186
Specifications
  • Description:
    Electrostatic motor
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Consists of a plastic cylinder placed on an insulating rod between two tip electrodes. When connecting a high-voltage source, a charge transport is obtained which causes the cylinder to rotate.
  • The following are included: rotor with bearings, rod with needle, tip electrodes with 4 mm contact pins (4 pcs.), Insulating rods with 4 mm contact sleeves (2 pcs.). Suitable accessory: influence machine