Tricarballylic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,2,3-Propantricarboxylic acid, Propane-1,2,3-tricarboxylic acid
CAS Number:99-14-9
:
Formula:C₆H₈O₆
MDL Number:MFCD00002723
Boiling Pt:266
Molecular Weight:176.13 g/mol
Melting Pt:154…159 °C
Merck Index:12,09752
EINECS:202-733-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Off-white to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:154 to 159 °C
- Titration with TBAH:≥98.5 %
Specification Test Results
Frequently Bought Together