N-Phenylmaleimide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:941-69-5
:
Formula:C₁₀H₇NO₂
MDL Number:MFCD00005502
Boiling Pt:162…163 °C (12 mmHg)
Molecular Weight:173.17 g/mol
Melting Pt:86…90 °C
ADR:6.1 III
Merck Index:12,07452
EINECS:213-382-0
UN:2811
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:86 to 91 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together