5,6-Dimethylbenzimidazole 99+%
: 5,6-Dimethyl-1H-benzimidazole, 5,6-Dimethyl-1H-benzo[d]imidazole
:582-60-5
:
:C₉H₁₀N₂
:MFCD00005603
:377
:146.19 g/mol
:203…206 °C
:13,03263
:209-488-1
- Environmentally Preferable:
- Appearance:Light beige Powder
- Infrared spectrum:Conforms
- Melting point:203 to 206 °C
- Titration with HClO4:≥99.0 %
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤20 ppm
- Sodium (Na):≤5000 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 %
Frequently Bought Together