Order Entry
Denmark
ContactUsLinkComponent
 
Nile red ≥95%
  AATB22190
 :  
undefined
Nile red ≥95%
  AATB22190
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315
  • Environmentally Preferable:

 

 

 : 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
 :
 :318.38 g/mol
 :GHS07
 :7385-67-3
 :C₂₀H₁₈N₂O₂
 :MFCD00011639
 :5 II