PCR plates, 384-well, Thermo-Fast® 384
PP. Designed for high throughput PCR, this 384-well plate is compatible with the majority of 384 block PCR and sequencing instruments. The full skirt and rigid plate deck minimise distortion during PCR making it suitable for use with robotic systems.
- Cut-away corners
- Fully skirted for compatibility with automated systems
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
40 µl maximum well volume, 25 µl working volume
Frequently Bought Together