Thermometer Temperature Indicating Label
Supplier: Brady
Certificates
About this item
Temperature indicating labels are a cost-effective solution for temperature checks during maintenance or for trouble spotting during research and development. The labels extremely low profile allows for application on components, on industrial and household applications such as electric pumps, rotating equipment, brakes and refrigerators, in cold chains and hot spaces or in areas where measuring temperature becomes difficult or impractical.
- Sub-surface printed polyester
- Gloss finish
- Cost effective
Application like cold temperature labelling, component and equipment labelling, facility identification, food and beverage process labelling, general identification, laboratory labelling, lean and 5S labelling, temperature indicating labelling.
Frequently Bought Together