Omnifix® Luer-Lock and Luer-Slip Syringes, Non sterile
Supplier: B. Braun
Certificates
About this item
Standard, fine dose and catheterfitting syringes.
- High visibility - transparent barrel with bold graduations allows for exact dosing of fluid
- Excellent plunger slide characteristics and secure backstop to prevent accidental plunger withdrawal
- Double seal stopper to prevent medication leakage
- Large, stable finger grips
- Minimal residual volume
- Components do not contain DEHP or natural rubber latex
Material:
Barrel - Polypropylene
Plunger - Polypropylene
Piston - Polyisoprene
Colour:
Barrel - Clear
Plunger - Clear
Piston - Black
Frequently Bought Together