Over-the-Spill Station
Supplier: Rubbermaid Commercial Products
Certificates
About this item
Dispensing station mounts on flat surfaces and has pads that soak up water and oil quickly and completely for an effective way to deal with spills.
- Slip-resistant pads quickly cover spills and alert pedestrians
- Each yellow pad is coded with universal caution symbol and 'Caution Wet Floor' message in English and Spanish
- Melt-blown polypropylene pads absorb water and oil
The Rubbermaid Commercial over-the-spill station is a dispensing station that mounts on flat surfaces and includes pad dispenser, 22 large pads, and fasteners.
Over-the-Spill® Tablet:
• Tablet mounts easily onto a floor sign or any flat surface
• Each medium size pad absorbs up to 0,34 L of liquid
• Pads resist slipping
• Perforated pads are easy to remove
Frequently Bought Together