Thermo Scientific™ TSX series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2 to 8 °C. The series V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
- Designed to safely store media, reagents, pharmaceuticals, vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 2 to 8 °C
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Redundant cooling offers additional peace of mind for your most precious samples
- Automated electronic passcode locking standard
- Datalogging with USB download functionality
- Designed for an ergonomic and comfortable experience
Unique to the TSX505, our dual refrigeration solution operates through the action of two synchronized solid-state heat pumps. The two solid-state heat pumps work to cool a combination of cold wall and forced air to help ensure that conditions are optimal for the most sensitive applications.
- Catalog No:
- HERATSX505SA
- HERATSX505GA
- Refrigerant:
- R744
- R744
- Display:
- Full color TFT-LCD, 4,3", 480×272
- Full color TFT-LCD, 4,3", 480×272
- Defrost:
- Automatic
- Automatic
- Alarm:
- Alarm
- Alarm
- Electrical:
- 100/240 V, 50/60 Hz
- 100/240 V, 50/60 Hz
- Door type:
- Solid
- Glass
- Plug:
- NEMA 5-15 P, more global types available
- NEMA 5-15 P, more global types available
- Weight:
- 53,98 kg
- 53,98 kg
- Shipping weight:
- 71 kg
- 71 kg