Order Entry
Denmark
ContactUsLinkComponent
Cole-Parmer® SH-200 See-Saw Rocking Shakers
Cole-Parmer® SH-200 See-Saw Rocking Shakers
Supplier:  Cole Parmer
Certificates
undefined
Cole-Parmer® SH-200 See-Saw Rocking Shakers
Supplier:  Cole Parmer
Specifications
  • Catalog No:
  • STUA51901-14
  • STUA51901-06
  • Model:
  • Mini
  • Large
  • Speed range:
  • 5 - 70 min⁻¹
  • 5 - 70 min⁻¹
  • Tilt angle:
  • W×D×H:
  • 275×335×155 mm
  • 390×450×165 mm
  • Platform W×D:
  • 235×235 mm
  • 355×355 mm
  • Display:
  • LED
  • LED
  • Max. load:
  • 3 kg
  • 10 kg
  • Timer:
  • 1 - 999 min
  • 1 - 999 min