PermaSleeve® Heat Shrink Wire and Cable Labels for M710 and BMP71
: Brady
PermaSleeve® PS heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use and permanence to create long-lasting, high-quality identification.
- 3:1 heat shrinkable sleeve fit snugly around wires for maximum insulation protection and permanent identification
- Fade-resistant and smudge-proof for superior aesthetic appearance
- Cold resistant and flame-retardant with an operating temperature of −55 to +135 °C (−67 to +275 °F)
- Matte finish
Applications include electrical labelling and wire and cable labelling.
A ribbon is required to print this material. Use part number M7-R4300.
: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202. RoHS compliant. UL recognised.
Frequently Bought Together