- Antibody type:Primary
- Antigen name:GALM/Galactose mutarotase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:251-342/342
- Form:Liquid
- Gene ID:130589
- Antigen synonyms:Galactose mutarotase|GALM_HUMAN|IBD1|Aldose 1-epimerase|Galm|Aldose 1 epimerase|Galactomutarotase|BLOCK25
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:38 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GALM/Galactose mutarotase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Galactose Mutarotase is a member of the aldose epimerase family and is involved in hexose metabolism. Through its catalytic activity, Galactose Mutarotase converts beta-aldose to alpha-aldose on several sugars, including D-glucose, L-arabinose and D-xylose. Found in the cytoplasm of most cells, Galactose Mutarotase plays a key role in galactose metabolism by catalyzing the conversion of beta-D-galactose to alpha-D-galactose. The enzyme contains two residues, Glu 304 and His 170, that are critical for catalysis, as well as His 96 and Asp 243, which are important for proper substrate recognition by the active site. No known diseases have been associated with mutations in the Galactose Mutarotase gene, although inhibition of Galactose Mutarotase activity could potentially be associated with a build-up of unmetabolized sugars during metabolism.
Type: Primary
Antigen: GALM/Galactose mutarotase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-342/342
Host: Rabbit
Isotype: IgG
Reactivity: Rat