Order Entry
Denmark
ContactUsLinkComponent
Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
Certificates
undefined
Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
Specifications
  • Connection type (port 1):
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock Plug
  • Male Rotating Luer Lock Plug
  • Male Rotating Luer Lock Plug
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Male Rotating Luer Lock
  • Connection type (port 2):
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • Barb
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 1
  • 1
  • 1
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA
  • FDA;USP
  • FDA;USP
  • FDA;USP