Order Entry
Denmark
ContactUsLinkComponent
Edge sealing compound, Safety-Walk™
  MMMASWKANTEN
 :  3M
undefined
Edge sealing compound, Safety-Walk™
  MMMASWKANTEN
 :  3M
  • Description:
    Safety-Walk™ edge sealing compound
  • Environmentally Preferable:
  • Pk:
    12
Safety-Walk™ Edge Sealing Compound is a liquid, solvent-based sealer that protects exposed edges of slip-resistant tapes and treads from excessive moisture or liquids.
  • Protect the exposed edges of slip-resistant tapes and treads
  • Keep surfaces safe from excessive moisture and liquids
Safety-Walk™ Edge Sealing Compound comes with a convenient tube applicator and seals approximately 22 to 30 linear metres.