Masterflex® Sanitary Clamp Fittings (Nylon), Avantor®
Compatible with metal and plastic Tri-Clamp® fittings.
- Excellent chemical resistance; low leachable characteristics
- Autoclavable
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.
- Environmentally Preferable:
- Model:
- 30508-29
- 30508-21
- 30508-27
- 30508-25
- 30508-23
- Sterilisation:
- Autoclave, Gamma
- Autoclave, Gamma
- Autoclave, Gamma
- Autoclave, Gamma
- Autoclave, Gamma
- Warranty:
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect
Frequently Bought Together