Wire Mesh Overscreens for Sanitary Strainers
Supplier: Avantor Fluid Handling
Certificates
About this item
Protect products and expensive process equipment from coarse or fine particulates.
- Mesh overscreens are available in a variety of mesh sizes to meet your specific application
- Overscreens are hemmed smooth with no stray wire ends
- Overscreens fit both in-line and side-inlet sanitary strainer assemblies
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
- Environmentally Preferable:
Specifications
- Filter type:
- Strainer
- Strainer
- Strainer
- Strainer
- Strainer
- Strainer
- Length:
- 27,62 cm (10,875")
- 27,62 cm (10,875")
- 27,62 cm (10,875")
- 27,62 cm (10,875")
- 27,62 cm (10,875")
- 27,62 cm (10,875")
- Ø:
- 7,62 cm (3")
- 7,62 cm (3")
- 7,62 cm (3")
- 7,62 cm (3")
- 7,62 cm (3")
- 7,62 cm (3")