Culture media, ready to use, Compact Dry
Ready to use dehydrated film media in rigid plastic with lid for cultivating and counting microorganisms in food, for monitoring product quality and potential contamination. These plates can be used to test raw material as well as finished products. Microorganisms are identified by the colours that are developed by chromogenic substrates and redox indicators.
- Ready to use - place 1 ml sample onto the plate and incubate
- Easy to read - colonies are pigmented with different colours
- Easy to store at room temperature - long shelf life
- Space saving and stackable
Dry TC, EC, CF,YM, XSA, SL, LS are AOAC approved
Dry TC, EC, CF, ETB, XSA, SL, LS are MicroVal approved
Dry TC, EC, CF, ETB, XSA, SL, LS are NordVal approved
European validation and certification alternative methods acc. to EN ISO 16140
- Environmentally Preferable:
Compact Dry Compact Dry Swabs Compact Dry-AQ Compact Dry-EC Compact Dry-ETB
Product Literature
More
IFU Compactdry AQ Instructions FR-EN IFU Compactdry CF Instructions FR-EN IFU Compactdry EC Instructions FR-EN IFU Compactdry ETB Instructions FR-EN IFU Compactdry ETC Instructions FR-EN
More
NISS54051-0TC-0040_TDS_EN
Datasheet
Swabs for Compact Dry
Frequently Bought Together