Storage drawers on wheels, IRIS
These drawer storage units are available with 3, 4, or 6 drawers, in large and extra large sizes to suit all your requirements and that you can use to store and organise your office and workspace.
- Transparent drawers make contents easily visible
- Hygienic smooth walls for easy cleaning
- Easy 360° glide castors for added mobility (can be taken away if wished)
- Stop system to keep drawers secure
- Colour: Clear drawer and black frame
Made of durable PP virgin plastic, these storage drawer units have a large capacity and are mounted on wheels to easily move the tower around a workspace. They have a strong frame and come with easy pull handles. The stop system keeps the drawers in the tower preventing them from sliding off.
Available with 3, 4 or 6 drawers, in large and extra large sizes to suit all your requirements. Their strong sturdy design brings you a heavy-duty storage drawer on wheels that you can use to store and organise your office and workspace.
Frequently Bought Together