About this item
Arium® pro Type I water purification systems are designed for sophisticated users to meet all their requirements for reagent grade water, needed both for critical applications and routine analyses. Different versions are available for reliable and optimal analytical results.
- Compact design, flexibility of use and simplicity of maintenance
- Innovative glass operator interface with touch function
- Service and alarm functions supported by graphic displays
- Conductivity measurement for feed and product water
- Integrated monitoring of water consumption
- RNases: <0,004 ng/ml; DNases: <0,024 pg/µ
The glass touch display, with its integrated keys and intuitive interface, guides users by menu prompts for ease of operation in producing reagent grade water. The system's various functions, such as volume and time dispensing, as well as continuously adjustable flow rate, give users the greatest possible flexibility in routine applications. PIN-protected service functions, Ethernet port and a wide choice of data storage options are features that users are bound to appreciate.
The systems are available in a selection of bench top, wall mounted and under bench units adapt optimally to users’ specific workplace requirements.
Exceeds the requirements of standards ASTM, NCCLS, ISO and USP.
Specifications
- Catalog No:
- 171-0648
- 171-0560
- 171-0561
- 171-0385
- 171-0384
- 171-0388
- 171-0394
- 171-0387
- 171-0382
- 171-0390
- 171-0557
- 171-0558
- Model:
- arium® pro DI
- arium® pro VF
- arium® pro VF
- arium® pro UV
- arium® pro UF
- arium® pro VF
- arium® pro DI
- arium® pro UV
- arium® pro UF
- arium® pro VF
- arium® pro UV
- arium® pro UV
- Flow rate:
- Up to 2 l/min
- Up to 1,7 l/min
- Up to 1,7 l/min
- Up to 2 l/min
- Up to 1,7 l/min
- Up to 1,7 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 1,7 l/min
- Up to 1,7 l/min
- Up to 2 l/min
- Up to 2 l/min
- TOC content:
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- 1 - 4 ppb at 100 ppb feed water
- Resistivity at +25 °C:
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- 18,2 MΩ.cm
- Particles:
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- <1/ml (@ 0,2 μm)
- Endotoxin:
- N/A
- <0,001 EU/ml
- <0,001 EU/ml
- N/A
- <0,001 EU/ml
- <0,001 EU/ml
- N/A
- N/A
- <0,001 EU/ml
- <0,001 EU/ml
- N/A
- N/A
- Microorganisms:
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- W×D×H:
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- 350×451×492 mm
- Operating weight with full tank:
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- ~27 kg
- Weight:
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
- ~17 kg
Specifications
Related Information
Sartorius arium® Water Purification Systems
Product Literature
Ultrapure Water for HPLC Analysis