3-(Trifluoromethyl)quinoxalin-2-ol
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Hydroxy-3-(trifluoromethyl)quinoxaline, 3-(Trifluoromethyl)quinoxalin-2(1H)-one
CAS Number:58457-64-0
:MFCD00275307
Formula:C₉H₅F₃N₂O
MDL Number:MFCD00243991
Molecular Weight:214.15 g/mol
Melting Pt:248…249 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together