3-(4-Fluorophenyl)-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-Amino-3-(4-fluorophenyl)-1H-pyrazole
CAS Number:72411-52-0
:MFCD10565813
Formula:C₉H₈FN₃
MDL Number:MFCD01023677
Molecular Weight:177.18 g/mol
Melting Pt:109…111 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together