5-Bromo-3-(difluoromethoxy)pyridin-2-amine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:947249-13-0
:
Formula:C₆H₅BrF₂N₂O
MDL Number:MFCD12405789
Molecular Weight:239.02 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together