Order Entry
Denmark
ContactUsLinkComponent
 
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%
Catalog # APOSPC19687-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%
Catalog # APOSPC19687-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate
 :
 :216.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:773873-95-3
Formula:C₉H₆F₂O₄
MDL Number:MFCD06203771