N-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-N-methylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-(1,3-Dimethyl-1H-pyrazol-5-yl)-N-methylmethanamine
CAS Number:852227-87-3
:
Formula:C₇H₁₃N₃
MDL Number:MFCD08700823
Molecular Weight:139.2 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together